Assay Details
| Gene Symbol: AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308584.1 | NP_001295513.1 | 5 - 6 | 2231 | 58 | |
| GenBank mRNA | AF105713.1 | - | 4 - 5 | 643 | 58 | |
| KF227907.1 | - | 5 - 6 | 2231 | 58 | ||
More Information
Set Membership: |
|

AR
Genomic Map