Assay Details
| Gene Symbol: KCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973100.1 | XP_027828901.1 | 6 - 7 | 1772 | 60 | |
| XM_027973102.1 | XP_027828903.1 | 5 - 6 | 1715 | 60 | ||
More Information
Set Membership: |
|

KCND3
Genomic Map