Assay Details
| Gene Symbol: STRADB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004826.4 | XP_004004875.1 | 9 - 10 | 1065 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STRADB| Gene Symbol: STRADB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004826.4 | XP_004004875.1 | 9 - 10 | 1065 | 64 | |
Set Membership: |
|