Assay Details
| Gene Symbol: EIF5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003164.3 | XP_004003213.1 | 3 - 4 | 371 | 92 | |
| XM_027958395.1 | XP_027814196.1 | 3 - 4 | 371 | 92 | ||
More Information
Set Membership: |
|

EIF5A2
Genomic Map