Assay Details
| Gene Symbol: RND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012956.3 | XP_004013005.2 | 2 - 3 | 206 | 71 | |
| XM_012186143.3 | XP_012041533.1 | 4 - 5 | 2595 | 71 | ||
| XM_012186144.3 | XP_012041534.1 | 4 - 5 | 389 | 71 | ||
| XM_027974470.1 | XP_027830271.1 | 4 - 5 | 452 | 71 | ||
More Information
Set Membership: |
|

RND2
Genomic Map