Assay Details
| Gene Symbol: MELTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957010.1 | XP_027812811.1 | 5 - 6 | 788 | 60 | |
| XM_027957019.1 | XP_027812820.1 | 5 - 6 | 788 | 60 | ||
More Information
Set Membership: |
|

MELTF
Genomic Map