Assay Details
Gene Symbol: SMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190016.3 | XP_012045406.2 | 4 - 5 | 1825 | 61 | |
XM_027978113.1 | XP_027833914.1 | 4 - 5 | 1760 | 61 | ||
XM_027978114.1 | XP_027833915.1 | 4 - 5 | 1801 | 61 | ||
XM_027978115.1 | XP_027833916.1 | 4 - 5 | 2109 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
