Assay Details
| Gene Symbol: SUPT16H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010368.4 | XP_004010417.1 | 19 - 20 | 2673 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SUPT16H| Gene Symbol: SUPT16H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010368.4 | XP_004010417.1 | 19 - 20 | 2673 | 66 | |
Set Membership: |
|