Assay Details
| Gene Symbol: GNA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961831.1 | XP_027817632.1 | 4 - 5 | 678 | 82 | |
| XM_027961832.1 | XP_027817633.1 | 3 - 4 | 642 | 82 | ||
More Information
Set Membership: |
|

GNA12
Genomic Map