Assay Details
| Gene Symbol: FOSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005788.3 | XP_004005837.1 | 2 - 3 | 2198 | 84 | |
| XM_027966637.1 | XP_027822438.1 | 2 - 3 | 1197 | 84 | ||
| XM_027966638.1 | XP_027822439.1 | 2 - 3 | 2195 | 84 | ||
More Information
Set Membership: |
|

FOSL2
Genomic Map