Assay Details
| Gene Symbol: NPSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007904.4 | XP_004007953.2 | 6 - 7 | 753 | 77 | |
| XM_027969108.1 | XP_027824909.1 | 6 - 7 | 753 | 77 | ||
| XM_027969109.1 | XP_027824910.1 | 4 - 5 | 555 | 77 | ||
More Information
Set Membership: |
|

NPSR1
Genomic Map