Assay Details
| Gene Symbol: RAD51B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097134.2 | XP_014952620.1 | 5 - 6 | 529 | 82 | |
| XM_015097135.2 | XP_014952621.1 | 5 - 6 | 609 | 82 | ||
| XM_027971941.1 | XP_027827742.1 | 5 - 6 | 529 | 82 | ||
| XM_027971942.1 | XP_027827743.1 | 5 - 6 | 529 | 82 | ||
| XM_027971943.1 | XP_027827744.1 | 5 - 6 | 529 | 82 | ||
More Information
Set Membership: |
|

RAD51B
Genomic Map