Assay Details
| Gene Symbol: LOC101123531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964894.1 | XP_027820695.1 | 11 - 12 | 2586 | 70 | |
| XM_027964895.1 | XP_027820696.1 | 11 - 12 | 2217 | 70 | ||
More Information
Set Membership: |
|

LOC101123531
Genomic Map