Assay Details
| Gene Symbol: HNRNPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177617.3 | XP_012033007.1 | 2 - 3 | 255 | 60 | |
| XM_012177618.3 | XP_012033008.1 | 2 - 3 | 203 | 60 | ||
| XM_012177619.2 | XP_012033009.1 | 2 - 3 | 255 | 60 | ||
| XM_015095739.2 | XP_014951225.1 | 2 - 3 | 255 | 60 | ||
| XM_027969281.1 | XP_027825082.1 | 2 - 3 | 256 | 60 | ||
| XM_027969282.1 | XP_027825083.1 | 2 - 3 | 202 | 60 | ||
| XM_027969283.1 | XP_027825084.1 | 2 - 3 | 206 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027969283.1 transcripts.
Set Membership: |
|

HNRNPH1
Genomic Map