Assay Details
Gene Symbol: ARCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016078.4 | XP_004016127.1 | 3 - 4 | 609 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
