Assay Details
| Gene Symbol: C1H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002545.4 | XP_004002594.1 | 3 - 4 | 530 | 72 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1H1orf189| Gene Symbol: C1H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002545.4 | XP_004002594.1 | 3 - 4 | 530 | 72 | |