Assay Details
Gene Symbol: DOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970029.1 | XP_027825830.1 | 4 - 5 | 1804 | 60 | |
XM_027970030.1 | XP_027825831.1 | 4 - 5 | 1788 | 60 | ||
XM_027970031.1 | XP_027825832.1 | 5 - 6 | 1104 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
