Assay Details
| Gene Symbol: LOC101105041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020042.4 | XP_004020091.2 | 8 - 9 | 725 | 97 | |
| XM_012102578.3 | XP_011957968.2 | 8 - 9 | 603 | 97 | ||
| XM_027960170.1 | XP_027815971.1 | 7 - 8 | 804 | 97 | ||
More Information
Set Membership: |
|

LOC101105041
Genomic Map