Assay Details
| Gene Symbol: LOC101117458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012100981.3 | XP_011956371.1 | 7 - 8 | 778 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117458| Gene Symbol: LOC101117458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012100981.3 | XP_011956371.1 | 7 - 8 | 778 | 79 | |
Set Membership: |
|