Assay Details
| Gene Symbol: FABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114667.1 | NP_001108139.1 | 1 - 2 | 71 | 93 | |
| GenBank mRNA | EU301804.1 | - | 1 - 2 | 71 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FABP4| Gene Symbol: FABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114667.1 | NP_001108139.1 | 1 - 2 | 71 | 93 | |
| GenBank mRNA | EU301804.1 | - | 1 - 2 | 71 | 93 | |