Assay Details
| Gene Symbol: LDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012101800.2 | XP_011957190.1 | 1 - 2 | 151 | 68 | |
| XM_027959817.1 | XP_027815618.1 | 2 - 3 | 314 | 68 | ||
| XM_027959818.1 | XP_027815619.1 | 2 - 3 | 243 | 68 | ||
| GenBank mRNA | AF233079.1 | - | 1 - 2 | 125 | 68 | |
More Information
Set Membership: |
|

LDHA
Genomic Map