Assay Details
| Gene Symbol: ELOVL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009618.4 | XP_004009667.1 | 3 - 4 | 353 | 74 | |
| XM_012179510.3 | XP_012034900.1 | 2 - 3 | 208 | 74 | ||
More Information
Set Membership: |
|

ELOVL6
Genomic Map