Assay Details
| Gene Symbol: SLC50A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002573.4 | XP_004002622.1 | 2 - 3 | 221 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC50A1| Gene Symbol: SLC50A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002573.4 | XP_004002622.1 | 2 - 3 | 221 | 59 | |
Set Membership: |
|