Assay Details
| Gene Symbol: TNNI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015426.4 | XP_004015475.1 | 7 - 8 | 711 | 62 | |
| GenBank mRNA | KX110055.1 | - | 7 - 8 | 702 | 62 | |
More Information
Set Membership: |
|

TNNI3
Genomic Map