Assay Details
| Gene Symbol: SLAMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040288.2 | NP_001035378.1 | 6 - 7 | 987 | 75 | |
| GenBank mRNA | DQ228866.1 | - | 6 - 7 | 991 | 75 | |
| MG669627.1 | - | 6 - 7 | 969 | 75 | ||
More Information
Set Membership: |
|

SLAMF1
Genomic Map