Assay Details
| Gene Symbol: SLC39A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127272.1 | NP_001120744.1 | 1 - 2 | 213 | 60 | |
| XM_012177524.3 | XP_012032914.1 | 4 - 5 | 638 | 60 | ||
| GenBank mRNA | EU626220.1 | - | 1 - 2 | 213 | 60 | |
More Information
Set Membership: |
|

SLC39A3
Genomic Map