Assay Details
| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009254.1 | NP_001009254.1 | 2 - 3 | 504 | 85 | |
| GenBank mRNA | AJ001048.1 | - | 2 - 3 | 504 | 85 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009254.1 | NP_001009254.1 | 2 - 3 | 504 | 85 | |
| GenBank mRNA | AJ001048.1 | - | 2 - 3 | 504 | 85 | |