Assay Details
| Gene Symbol: RAB8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161860.1 | NP_001155332.1 | 1 - 2 | 125 | 78 | |
| GenBank mRNA | FJ943968.1 | - | 1 - 2 | 125 | 78 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB8A| Gene Symbol: RAB8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161860.1 | NP_001155332.1 | 1 - 2 | 125 | 78 | |
| GenBank mRNA | FJ943968.1 | - | 1 - 2 | 125 | 78 | |