Assay Details
| Gene Symbol: FGF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009235.2 | NP_001009235.2 | 2 - 3 | 384 | 152 | |
| XM_012180603.3 | XP_012035993.1 | 3 - 4 | 922 | 152 | ||
| GenBank mRNA | EU626202.1 | - | 2 - 3 | 384 | 152 | |
| Z46236.1 | - | 421 | 152 | |||
More Information
Set Membership: |
|

FGF7
Genomic Map