Assay Details
| Gene Symbol: GGTA2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009764.1 | NP_001009764.1 | 3 - 4 | 184 | 145 | |
| XM_012159476.3 | XP_012014866.1 | 5 - 6 | 380 | 145 | ||
| XM_027965748.1 | XP_027821549.1 | 5 - 6 | 809 | 145 | ||
| XM_027965749.1 | XP_027821550.1 | 5 - 6 | 586 | 145 | ||
| XM_027965750.1 | XP_027821551.1 | 6 - 7 | 681 | 145 | ||
| GenBank mRNA | AY447039.1 | - | 3 - 4 | 184 | 145 | |
More Information
Set Membership: |
|

GGTA2P
Genomic Map