Assay Details
| Gene Symbol: GGTA2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009764.1 | NP_001009764.1 | 5 - 6 | 426 | 95 | |
| XM_012159476.3 | XP_012014866.1 | 7 - 8 | 622 | 95 | ||
| XM_027965748.1 | XP_027821549.1 | 7 - 8 | 1051 | 95 | ||
| XM_027965749.1 | XP_027821550.1 | 7 - 8 | 828 | 95 | ||
| XM_027965750.1 | XP_027821551.1 | 8 - 9 | 923 | 95 | ||
| GenBank mRNA | AY447039.1 | - | 5 - 6 | 426 | 95 | |
More Information
Set Membership: |
|

GGTA2P
Genomic Map