Assay Details
| Gene Symbol: CATHL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009772.1 | NP_001009772.1 | 3 - 4 | 380 | 108 | |
| XM_027957553.1 | XP_027813354.1 | 3 - 4 | 383 | 108 | ||
| GenBank mRNA | AB974690.1 | - | 3 - 4 | 380 | 108 | |
| L46853.1 | - | 3 - 4 | 380 | 108 | ||
| Gene Symbol: LOC114109381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957969.1 | XP_027813770.1 | 3 - 4 | 383 | 108 | |
| XM_027957970.1 | XP_027813771.1 | 3 - 4 | 380 | 108 | ||
| Gene Symbol: LOC114109382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957971.1 | XP_027813772.1 | 3 - 4 | 383 | 108 | |
| XM_027957972.1 | XP_027813773.1 | 3 - 4 | 380 | 108 | ||
More Information
Set Membership: |
|

CATHL1B
Genomic Map