Assay Details
| Gene Symbol: IL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009783.1 | NP_001009783.1 | 1 - 2 | 179 | 73 | |
| GenBank mRNA | AY575612.1 | - | 1 - 2 | 36 | 73 | |
| U35038.1 | - | 1 - 2 | 179 | 73 | ||
More Information
Set Membership: |
|

IL5
Genomic Map