Assay Details
| Gene Symbol: RORB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129737.1 | NP_001123209.1 | 4 - 5 | 754 | 68 | |
| GenBank mRNA | EF494242.1 | - | 4 - 5 | 754 | 68 | |
More Information
Set Membership: |
|

RORB
Genomic Map