Assay Details
| Gene Symbol: ARFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162549.1 | NP_001156021.1 | 4 - 5 | 542 | 67 | |
| XM_012095242.3 | XP_011950632.1 | 5 - 6 | 674 | 67 | ||
| XM_012095243.1 | XP_011950633.1 | 2 - 3 | 287 | 67 | ||
| XM_027978975.1 | XP_027834776.1 | 5 - 6 | 642 | 67 | ||
| GenBank mRNA | FJ969418.1 | - | 4 - 5 | 542 | 67 | |
More Information
Set Membership: |
|

ARFIP2
Genomic Map