Assay Details
| Gene Symbol: TIMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166187.1 | NP_001159659.1 | 1 - 2 | 123 | 60 | |
| GenBank mRNA | GQ221039.1 | - | 1 - 2 | 123 | 60 | |
More Information
Set Membership: |
|

TIMP3
Genomic Map