Assay Details
| Gene Symbol: THRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190391.1 | NP_001177320.1 | 5 - 6 | 738 | 53 | |
| GenBank mRNA | HM055898.1 | - | 5 - 6 | 738 | 53 | |
| Z68307.1 | - | 7 - 8 | 1098 | 53 | ||
More Information
Set Membership: |
|

THRB
Genomic Map