Assay Details
| Gene Symbol: MYOZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199795.1 | NP_001186724.1 | 4 - 5 | 344 | 59 | |
| GenBank mRNA | HM628580.1 | - | 4 - 5 | 344 | 59 | |
More Information
Set Membership: |
|

MYOZ3
Genomic Map