Assay Details
| Gene Symbol: CXCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009191.1 | NP_001009191.1 | 2 - 3 | 239 | 85 | |
| GenBank mRNA | AB070717.1 | - | 2 - 3 | 239 | 85 | |
| HM017954.1 | - | 2 - 3 | 180 | 85 | ||
More Information
Set Membership: |
|

CXCL10
Genomic Map