Assay Details
| Gene Symbol: CLN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082595.1 | NP_001076064.1 | 3 - 4 | 568 | 80 | |
| GenBank mRNA | EF204938.1 | - | 3 - 4 | 568 | 80 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CLN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082595.1 | NP_001076064.1 | 3 - 4 | 568 | 80 | |
| GenBank mRNA | EF204938.1 | - | 3 - 4 | 568 | 80 | |