Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246236.1 | NP_001233165.1 | 3 - 4 | 704 | 69 | |
| XM_027969179.1 | XP_027824980.1 | 4 - 5 | 923 | 69 | ||
| XM_027969180.1 | XP_027824981.1 | 4 - 5 | 888 | 69 | ||
| GenBank mRNA | FN908185.1 | - | 3 - 4 | 704 | 69 | |
More Information
Set Membership: |
|

ZNF496
Genomic Map