Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246236.1 | NP_001233165.1 | 5 - 6 | 960 | 93 | |
| XM_027969179.1 | XP_027824980.1 | 6 - 7 | 1179 | 93 | ||
| XM_027969180.1 | XP_027824981.1 | 6 - 7 | 1144 | 93 | ||
| GenBank mRNA | FN908185.1 | - | 5 - 6 | 960 | 93 | |
| FN908186.1 | - | 2 - 3 | 100 | 93 | ||
More Information
Set Membership: |
|

ZNF496
Genomic Map