Assay Details
| Gene Symbol: PDXK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009220.1 | NP_001009220.1 | 8 - 9 | 791 | 56 | |
| XM_027966611.1 | XP_027822412.1 | 10 - 11 | 967 | 56 | ||
| GenBank mRNA | AF125374.1 | - | 8 - 9 | 791 | 56 | |
More Information
Set Membership: |
|

PDXK
Genomic Map