Assay Details
| Gene Symbol: PRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009306.1 | NP_001009306.1 | 2 - 3 | 306 | 70 | |
| GenBank mRNA | M27057.1 | - | 2 - 3 | 306 | 70 | |
| X13483.1 | - | 238 | 70 | |||
More Information
Set Membership: |
|

PRL
Genomic Map