Assay Details
| Gene Symbol: CSN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009378.1 | NP_001009378.1 | 3 - 4 | 163 | 73 | |
| GenBank mRNA | KC963133.1 | - | 3 - 4 | 140 | 73 | |
| KC963134.1 | - | 3 - 4 | 140 | 73 | ||
| X51822.1 | - | 163 | 73 | |||
More Information
Set Membership: |
|

CSN3
Genomic Map