Assay Details
| Gene Symbol: RPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280688.1 | NP_001267617.1 | 3 - 4 | 340 | 99 | |
| GenBank mRNA | JX534531.1 | - | 3 - 4 | 340 | 99 | |
More Information
Set Membership: |
|

RPS17
Genomic Map