Assay Details
| Gene Symbol: IZUMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318079.1 | NP_001305008.1 | 1 - 2 | 277 | 64 | |
| GenBank mRNA | KR020834.1 | - | 1 - 2 | 277 | 64 | |
More Information
Set Membership: |
|

IZUMO2
Genomic Map