Assay Details
| Gene Symbol: SFXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126350.1 | NP_001119822.1 | 4 - 5 | 512 | 60 | |
| XM_027971424.1 | XP_027827225.1 | 5 - 6 | 603 | 60 | ||
| XM_027971425.1 | XP_027827226.1 | 5 - 6 | 591 | 60 | ||
| GenBank mRNA | EU583468.1 | - | 4 - 5 | 512 | 60 | |
More Information
Set Membership: |
|

SFXN1
Genomic Map