Assay Details
| Gene Symbol: HSD3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135932.1 | NP_001129404.1 | 2 - 3 | 315 | 95 | |
| XM_012183658.2 | XP_012039048.1 | 3 - 4 | 502 | 95 | ||
| GenBank mRNA | FJ007375.1 | - | 2 - 3 | 315 | 95 | |
More Information
Set Membership: |
|

HSD3B1
Genomic Map