Assay Details
| Gene Symbol: TP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009403.1 | NP_001009403.1 | 2 - 3 | 205 | 67 | |
| GenBank mRNA | FJ855223.1 | - | 1 - 2 | 79 | 67 | |
| X81705.1 | - | 205 | 67 | |||
More Information
Set Membership: |
|

TP53
Genomic Map